Diphenylsilanediol
Catalog # CHMPSILS06653.
Supplier: CHEMPUR
CAS Number:
Specifications
- Pk:1
Specifications
About this item
:
:216.31 g/mol
Boiling Pt:377 °C (1013 hPa)
Melting Pt:174 °C
Density:1.25 g/cm³ (20 °C)
EINECS:213-427-4
UN:1325
ADR:4.1,III
CAS Number:947-42-2
Formula:C₁₂H₁₂O₂Si
MDL Number:MFCD00002101