Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(Dichloromethyl)trimethylsilane
(Dichloromethyl)trimethylsilane
(Dichloromethyl)trimethylsilane
(Dichloromethyl)trimethylsilane
Supplier:
CHEMPUR SILANE
Certificates
Expand image
(Dichloromethyl)trimethylsilane
Supplier:
CHEMPUR SILANE
About this item
CAS Number:
5926-38-5
:
Formula:
Cl₂HCSi(CH₃)₃
:
157.11 g/mol
Boiling Pt:
133 °C (1013 hPa)
Density:
1.0395 g/cm³ (25 °C)
EINECS:
227-658-3