Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(Chloromethyl)trichlorosilane
(Chloromethyl)trichlorosilane
(Chloromethyl)trichlorosilane
(Chloromethyl)trichlorosilane
Supplier:
CHEMPUR SILANE
Certificates
Expand image
(Chloromethyl)trichlorosilane
Supplier:
CHEMPUR SILANE
About this item
CAS Number:
1558-25-4
:
Formula:
ClH₂CSiCl₃
MDL Number:
MFCD00000873
:
183.92 g/mol
Boiling Pt:
117 °C (1013 hPa)
Density:
1.442 g/cm³ (20 °C)
EINECS:
216-316-9