Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bis(dimethylamino)diethylsilane
Bis(dimethylamino)diethylsilane
Bis(dimethylamino)diethylsilane
Bis(dimethylamino)diethylsilane
Supplier:
CHEMPUR SILANE
Certificates
Expand image
Bis(dimethylamino)diethylsilane
Supplier:
CHEMPUR SILANE
About this item
CAS Number:
33287-52-4
:
Formula:
C₈H₂₂N₂Si
:
174.36 g/mol