Tetraphenylcyclopentadienone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:479-33-4
:
Formula:C₂₉H₂₀O
MDL Number:MFCD00001407
Melting Pt:219…222 °C
Density:1.233 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:384.48 g/mol
EINECS:207-530-3