Squalene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:111-02-4
:
Formula:C₃₀H₅₀
MDL Number:MFCD00008912
:410.73 g/mol
Merck Index:13,08847
Boiling Pt:429 °C (1013 hPa)
Melting Pt:–5 °C
Density:0.86 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Flash Pt:200 °C
EINECS:203-826-1