Benzoic acid methyl ester, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:93-58-3
:
Formula:C₆H₅COOCH₃
MDL Number:MFCD00008421
:136.15 g/mol
Merck Index:13,06052
Boiling Pt:199.6 °C (1013 hPa)
Melting Pt:–12 °C
Density:1.0888 g/cm³ (20 °C)
Flash Pt:83 °C
EINECS:202-259-7
:GHS07
Storage Temperature:Ambient