Furfuryl mercaptan, Sigma-Aldrich®
Catalog # SIALF20408-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:114.17 g/mol
Boiling Pt:155 °C (1013 hPa)
Density:1.1339 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:51 °C
EINECS:202-628-2
CAS Number:98-02-2
Formula:C₅H₆OS
MDL Number:MFCD00003254