3,5-Dimethylisoxazole, Sigma-Aldrich®
Catalog # SIALD167509-25ML
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mL
- Seq:2315
Specifications
About this item
:
:97.12 g/mol
Boiling Pt:142 °C (1013 hPa)
Density:0.98 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:206-100-2
CAS Number:300-87-8
Formula:C₅H₇NO
MDL Number:MFCD00003156