Order Entry
Germany
Orders LinkContactUsLinkComponent
Cycloheximide, Sigma-Aldrich®

Cycloheximide, Sigma-Aldrich®

Toxic
Health
Environment
Supplier:   Merck
Cycloheximide, Sigma-Aldrich®
Supplier:  Merck
Certificates
Cycloheximide, Sigma-Aldrich®
Cycloheximide, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
Synonyms:  Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
  • CAS Number:66-81-9
  • Formula:C₁₅H₂₃NO₄
  • MDL Number:MFCD00082346
  • Molecular Weight:281.35 g/mol
  • Storage Temperature:Ambient
  • ADR:6.1,I
  •  :M-Clarity™ quality level = MQ100
  • EINECS:200-636-0
  • UN:2811
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 118,00 €
    118,00 €
    1 * 1 g
       118,00 €

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 463,00 €
    463,00 €
    1 * 5 g
       463,00 €