1-Naphtholbenzeine, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:145-50-6
:6948-88-5
Formula:C₂₇H₁₈O₂
MDL Number:MFCD00078492
:374.44 g/mol
Melting Pt:243…245 °C
:M-Clarity™ quality level = MQ100
EINECS:205-656-3
:GHS07
Storage Temperature:Ambient