Dysprosium(III) oxide, Sigma-Aldrich®
Catalog # SIAL637289-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
Merck Index:14,03482
Melting Pt:∼2400 °C
Density:7.81 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:373 g/mol
EINECS:215-164-0
CAS Number:1308-87-8
Formula:Dy₂O₃
MDL Number:MFCD00010986