Order Entry
Germany
ContactUsLinkComponent
 
3-(4-Fluorophenyl)propionic acid, Sigma-Aldrich®
Catalog # SIAL560502-1G
Supplier:  Merck
CAS Number:  
3-(4-Fluorophenyl)propionic acid, Sigma-Aldrich®
Catalog # SIAL560502-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :168.17 g/mol
 :M-Clarity™ quality level = MQ100
CAS Number:459-31-4
Formula:C₉H₉FO₂
MDL Number:MFCD00060327