4-Fluorobenzyl mercaptan, Sigma-Aldrich®
Catalog # SIAL539155-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:142.2 g/mol
Boiling Pt:37 °C (0.1 mmHg)
Density:1.157 g/cm³ (20 °C)
ADR:6.1,III
:M-Clarity™ quality level = MQ100
EINECS:240-031-9
UN:2810
CAS Number:15894-04-9
Formula:C₇H₇FS
MDL Number:MFCD00040724