1-Methyluracil, Sigma-Aldrich®
Catalog # SIAL293768-500MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:126.11 g/mol
:M-Clarity™ quality level = MQ200
CAS Number:615-77-0
Formula:C₅H₆N₂O₂
MDL Number:MFCD00038666