Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cesium hydrogen carbonate
Cesium hydrogen carbonate, Sigma-Aldrich®
Cesium hydrogen carbonate, Sigma-Aldrich®
Cesium hydrogen carbonate, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Cesium hydrogen carbonate, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
15519-28-5
:
29703-01-3
Formula:
CHCsO₃
MDL Number:
MFCD00010961
:
193.92 g/mol
:
M-Clarity™ quality level = MQ100
EINECS:
239-554-5