
Maleimide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:541-59-3
:
Formula:C₄H₃NO₂
MDL Number:MFCD00005494
:97.07 g/mol
Boiling Pt:102…105 °C (55 mmHg)
Melting Pt:90…93 °C
Density:1.2493 g/cm³ (106 °C)
EINECS:208-787-4
ADR:8,III
:M-Clarity™ quality level = MQ100
UN:2923
:GHS05