4-Methoxybenzyl mercaptan, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6258-60-2
:
Formula:C₈H₁₀OS
MDL Number:MFCD00004871
:154.23
Boiling Pt:90…95 °C (0.67 hPa)
Density:1.11 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Refrigerator
EINECS:228-393-6