1,1,1-Trichloro-2-methyl-2-propanol hemihydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6001-64-5
:
Formula:C₈H₁₆Cl₆O₃
MDL Number:MFCD02179352
:372.93 g/mol
Boiling Pt:167 °C (1013 hPa)
Melting Pt:78 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-317-6