Neutral red
Glass bottle for solids, 25 g
SERA30305.01
Marketsource
: Serva
: N2,N2,7-Trimethylphenazine-2,8-diamine hydrochloride, C.I. 50040, 3-Amino-7-dimethyl amino-2-methylphenazine hydrochloride, N8,N8,3-Trimethyl-2,8-phenazinediamine monohydrochloride, Toluylene red, Basic Red 5
- :553-24-2
- :C₁₅H₁₇ClN₄
- :MFCD00012651
- :288.78 g/mol
- :12,06571
- :290 °C
- :Ambient
- :209-035-8
- Pack type:Glass bottle for solids
- Pk:25 g