
2-(4-Iodophenyl)-3-(4-nitrophenyl)-5-phenyltetrazolium chloride
Supplier: Serva
Certificates
About this item
CAS Number:146-68-9
:
Formula:C₁₉H₁₃ClIN₅O₂
MDL Number:MFCD00011961
:505.7 g/mol
Melting Pt:227…229 °C
EINECS:205-676-2
:GHS08
Storage Temperature:Refrigerator
Frequently Bought Together