Order Entry
Germany
ContactUsLinkComponent
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate
Catalog # PROOMM0535.00
CAS Number:  
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate
Catalog # PROOMM0535.00
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :372.93 g/mol
Boiling Pt:167 °C (1013 hPa)
Melting Pt:78 °C
Storage Temperature:Ambient
EINECS:200-317-6
CAS Number:6001-64-5
Formula:C₈H₁₆Cl₆O₃
MDL Number:MFCD02179352