Charcoal activated, powder, AnalaR NORMAPUR® analytical reagent
Catalog # 87126.230
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:250 g
- Seq:3545
Specifications
Specification Test Results
- Decolourising value:Min. 20 ml
- Heavy metals (as Pb):Max. 20 ppm
- Ignition residue (SO4):Max. 7 %
- Loss on drying (120°C):Max. 8 %
- Solubility in diluted HCl:Max. 10.0 %
- Solubility in ethanol:Max. 0.2 %
- Solubility in water:Max. 2.0 %
- Soluble chlorides:Max. 0.1 %
- Soluble sulphates:Max. 100 ppm
- Ca (Calcium):Max. 1 %
- Fe (Iron):Max. 0.03 %
- Zn (Zinc):Max. 10 ppm
Specification Test Results
About this item
:
:12.01 g/mol
Merck Index:13,01818
Melting Pt:3550 °C
Density:2 g/cm³ (20 °C)
EINECS:231-153-3
UN:1362
ADR:4.2,III
REACH:01-2119488894-16
CAS Number:64365-11-3
Formula:C
MDL Number:MFCD00133992