Order Entry
Germany
ContactUsLinkComponent
Thioacetamide
Thioacetamide
Catalog # 28600.183
Supplier:  VWR Chemicals
CAS Number:  
undefined
Thioacetamide
Catalog # 28600.183
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :75.13 g/mol
Merck Index:13,09391
Melting Pt:113…114 °C
Density:1.336 g/cm³ (20 °C)
EINECS:200-541-4
 :GHS08
 :GHS07
CAS Number:62-55-5
Formula:CH₃CSNH₂
MDL Number:MFCD00008070