Nickel sulphate heptahydrate
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:10101-98-1
:
Formula:NiO₄S·7H₂O
MDL Number:MFCD00149814
Melting Pt:99 °C
Density:∼2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:232-104-9
UN:3077
ADR:9,III
:280.86 g/mol
:GHS08
:GHS07
:GHS09