Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl m-toluate
Methyl-m-toluate
Methyl-m-toluate
Methyl-m-toluate
Catalog # MOLE11372927-100G
Supplier:
Molekula
CAS Number:
Expand image
Methyl-m-toluate
Catalog # MOLE11372927-100G
Supplier:
Molekula
CAS Number:
Specifications
Pk:
100 g
Seq:
3401
Specifications
About this item
Product Details
:
:
150.18
EINECS:
202-753-2
CAS Number:
99-36-5
Formula:
C₉H₁₀O₂
MDL Number:
MFCD00008436
Product Details