2-(4-Bromophenyl)ethanol
Glass bottle for solids, 5 g
Catalog # MOLE41183694-5G
Marketsource
Supplier: Molekula
2-(4-Bromophenyl)ethanol
Catalog # MOLE41183694-5G
Supplier: Molekula
Marketsource
CAS Number: 4654-39-1
Jump to:
Product Details & Documents
Synonyms: 4-Bromophenethyl alcohol
- CAS Number:4654-39-1
- Formula:C₈H₉BrO
- MDL Number:MFCD00002897
- Molecular Weight:201.06 g/mol
- Boiling Pt:275 °C (1013 hPa)
- Melting Pt:36…38 °C
- Density:1,4678 g/cm³ (28.5 °C)
- EINECS:225-093-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



