Order Entry
Germany
ContactUsLinkComponent
1-(2-Pyridyl)piperazine
1-(2-Pyridyl)piperazine
Catalog # MOLE53917698-25G
Supplier:  Molekula
CAS Number:  
1-(2-Pyridyl)piperazine
Catalog # MOLE53917698-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :163.22 g/mol
Boiling Pt:293 °C (1013 hPa)
Melting Pt:85…87 °C
Density:1,07 g/cm³ (20 °C)
EINECS:252-220-3
CAS Number:34803-66-2
Formula:C₉H₁₃N₃
MDL Number:MFCD00006216