Order Entry
Germany
ContactUsLinkComponent
Tetrabutylammonium periodate
Tetrabutylammonium periodate
Catalog # MOLE90030525-25G
Supplier:  Molekula
CAS Number:  
Tetrabutylammonium periodate
Catalog # MOLE90030525-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :433.37 g/mol
Melting Pt:175 °C
EINECS:265-614-5
UN:1479
ADR:5.1,III
CAS Number:65201-77-6
Formula:C₁₆H₃₆INO₄