Order Entry
Germany
Orders LinkContactUsLinkComponent
 

2-Deoxy-D(-)-ribose

Glass bottle for solids, 25 g
Catalog # MOLE12691995-25G
Marketsource
Supplier:   Molekula
2-Deoxy-D(-)-ribose
Catalog # MOLE12691995-25G
Supplier:  Molekula
Marketsource
CAS Number:  533-67-5
2-Deoxy-D(-)-ribose

Glass bottle for solids, 25 g

CAS Number:  533-67-5
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  2-Deoxy-D(-)-erythropentose, Thyminose, 2-Deoxy-D(-)-arabinose, 2'-Deoxy-D-Ribose, (3S,4R)-3,4,5-Trihydroxypentanal
  • CAS Number:533-67-5
  • Formula:C₅H₁₀O₄
  • MDL Number:MFCD00135904
  • Molecular Weight:134.13 g/mol
  • Merck Index:13,02925
  • Boiling Pt:292 °C (1013 hPa)
  • Melting Pt:95…97 °C
  • Storage Temperature:Ambient
  • EINECS:208-573-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications