Order Entry
Germany
ContactUsLinkComponent
 
trans-1,2-Cyclohexanedicarboxylic acid for synthesis, Sigma-Aldrich®
Catalog # 8.41088.0005
Supplier:  Merck
CAS Number:  
trans-1,2-Cyclohexanedicarboxylic acid for synthesis, Sigma-Aldrich®
Catalog # 8.41088.0005
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Melting Point:
    220 - 221 °C
  • Molar Mass:
    172.18 g/mol
  • Synonyms:
    trans-Hexahydrophthalic acid
  • Hill Formula:
    C8H12O4

Specification Test Results

About this item

 :
 :172.18 g/mol
Boiling Pt:339 °C (1013 hPa)
Melting Pt:220…221 °C
 :M-Clarity™ quality level = MQ200
EINECS:218-975-8
CAS Number:2305-32-0
Formula:C₈H₁₂O₄
MDL Number:MFCD00001464