Order Entry
Germany
ContactUsLinkComponent
N,N-Dimethylethylamine, Sigma-Aldrich®
Flammable
Corrosive
Irritant
N,N-Dimethylethylamine, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Corrosive
Irritant
N,N-Dimethylethylamine, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    (20 °C) miscible
  • Melting Point:
    -140 °C
  • Molar Mass:
    73.14 g/mol
  • Boiling Point:
    36.5 °C (1013 hPa)
  • Vapor Pressure:
    527 hPa (20 °C)
  • Flash Point:
    -45.5 °C
  • Explosion Limit:
    0.9 - 11.2 %(V)
  • Density:
    0.675 g/cm3 (20 °C)
  • pH:
    12.0 (50 g/l, H2O, 20 °C)
  • Ignition Point:
    190 °C

Specification Test Results

Related Information

Frequently Bought Together