Order Entry
Germany
ContactUsLinkComponent
Dibenzothiophene, Sigma-Aldrich®
Environment
Dibenzothiophene, Sigma-Aldrich®
Catalog # 8.20409.0025
Supplier:  Merck
CAS Number:  
Environment
Dibenzothiophene, Sigma-Aldrich®
Catalog # 8.20409.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Solubility:
    0.0015 g/l (25 °C) (Lit.)
  • Melting Point:
    95 - 98 °C
  • Molar Mass:
    184.25 g/mol
  • Boiling Point:
    331 - 333 °C (1013 hPa)
  • Flash Point:
    170 °C
  • pH:
    7 (50 g/l, H2O, 20 °C) (slurry)
  • Ignition Point:
    450 °C

Specification Test Results

About this item

 :
 :184.26 g/mol
Boiling Pt:332 °C (1013 hPa)
Melting Pt:95…98 °C
Density:1.1295 g/cm³ (125 °C)
Flash Pt:170 °C
Storage Temperature:Ambient
EINECS:205-072-9
UN:3077
ADR:9,III
 :M-Clarity™ quality level = MQ200
 :GHS09
CAS Number:132-65-0
Formula:C₁₂H₈S
MDL Number:MFCD00004969