Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-82 °C
- Molar Mass:75.11 g/mol
- Boiling Point:95 °C (1013 hPa)
- Flash Point:12 °C
- Refractive Index:1.4594 (20 °C, 589 nm)
- Explosion Limit:3 - 16.10 %(V)
- Density:0.87 g/cm3 (20 °C)
- pH:12 (100 g/l, H2O, 20 °C)
- Ignition Point:285 °C
- Hazard Class:3
- Hazard Class Description:Flammable Liquid