1,3-Diethyl-2-thiourea, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:105-55-5
:
Formula:C₅H₁₂N₂S
MDL Number:MFCD00004925
:132.23 g/mol
Boiling Pt:169 °C (1013 hPa)
Melting Pt:75…78 °C
Density:1.118 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:186 °C
EINECS:203-308-5
:GHS05
:GHS08
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:38 g/l (20 °C)
- Melting Point:75 - 78 °C
- Molar Mass:132.22 g/mol
- Bulk Density:735 kg/m3
- Flash Point:186 °C
- Density:1.12 g/cm3 (20 °C)
- pH:7 (38 g/l, H2O, 20 °C)