Order Entry
Germany
ContactUsLinkComponent
Nitric acid 69%, Suprapur®, Supelco®
Nitric acid 69%, Suprapur®, Supelco®
Catalog # 1.15187.1000
Supplier:  Merck
CAS Number:  
Nitric acid 69%, Suprapur®, Supelco®
Catalog # 1.15187.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :63.01 g/mol
Merck Index:13,06608
Boiling Pt:120.5 °C (1013 hPa)
Melting Pt:–42 °C
Density:1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8 (5.1),
 :M-Clarity™ quality level = MQ100
 :GHS03
 :GHS06
 :GHS05
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349