1,1,1-Trichloro-2-methyl-2-propanol hemihydrate Ph. Eur., BP, JP, NF, SAFC®
Catalog # 1.01005.1000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:372.93 g/mol
Boiling Pt:167 °C (1013 hPa)
Melting Pt:78 °C
EINECS:200-317-6
:GHS07
Storage Temperature:Ambient
CAS Number:6001-64-5
Formula:C₈H₁₆Cl₆O₃
MDL Number:MFCD02179352