1-(2,5-Dichlorophenyl)-2-thiourea
Catalog # MENASR470.50
Supplier: MENAI
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:221.11 g/mol
Melting Pt:320 °C
UN:2811
ADR:6.1,III
CAS Number:4949-85-3
Formula:(C₆H₃Cl₂)NC(S)NH₂
MDL Number:MFCD00041154