Order Entry
Germany
Orders LinkContactUsLinkComponent
Ethanol absolute analytical reagent

Ethanol absolute analytical reagent

Flammable
   Sustainable Options Available
Supplier:   Bernd Kraft
Ethanol absolute analytical reagent
Supplier:  Bernd Kraft
   Sustainable Options Available
Certificates

7 Options Available Below

Product Details & Documents
Synonyms:  Ethyl alcohol
  • CAS Number:64-17-5
  • Formula:C₂H₅OH
  • MDL Number:MFCD00003568
  • Molecular Weight:46.07 g/mol
  • Merck Index:12,03806
  • Boiling Pt:78.3 °C (1013 hPa)
  • Melting Pt:–117 °C
  • Density:0.7895 g/cm³ (20 °C)
  • Flash Pt:12 °C
  • Storage Temperature:Ambient
  • EINECS:200-578-6
  • UN:1170
  • ADR:3,II
  • REACH:01-2119457610-43

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • KRAF07227.2700
  • KRAF07227.4000
  • KRAF07227.4700
  • KRAF07227.3000
  • KRAF07227.3700
  • KRAF07227.5300
  • KRAF07227.6700
  • Environmentally Preferable:
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Safer Human and Environmental Health
  • Environmental certification:
  • EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
  • EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
  • EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
  • EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
  • EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
  • EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
  • EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    KRAF07227.2700
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
      Safer Human and Environmental Health
  • Glass bottle
    2.500 mL
    Log in for availability
    Log in for availability
    1 * 2.500 mL
    1 * 2.500 mL - 229,00 €
    229,00 €
    1 * 2.500 mL
       229,00 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    KRAF07227.4000
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
      Safer Human and Environmental Health
  • Glass bottle
    2.500 mL
    Log in for availability
    Log in for availability
    1 * 2.500 mL
    1 * 2.500 mL - 232,00 €
    232,00 €
    1 * 2.500 mL
       232,00 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    KRAF07227.4700
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
      Safer Human and Environmental Health
  • Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 115,00 €
    115,00 €
    1 * 1 L
       115,00 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    KRAF07227.3000
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
      Safer Human and Environmental Health
  • Plastic bottle
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 117,00 €
    117,00 €
    1 * 1 L
       117,00 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    KRAF07227.3700
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
      Safer Human and Environmental Health
  • Plastic bottle
    5 L
    Log in for availability
    Log in for availability
    1 * 5 L
    1 * 5 L - 381,00 €
    381,00 €
    1 * 5 L
       381,00 €

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    KRAF07227.5300
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
      Safer Human and Environmental Health
  • Plastic bottle
    10 L
    Log in for availability
    Log in for availability
    1 * 10 L
    1 * 10 L

    Specifications:
      Safer Human and Environmental Health
    More
    Specifications
    Cat. No.
    KRAF07227.6700
    Environmentally Preferable
    Safer Human and Environmental Health
    Environmental certification
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
      Safer Human and Environmental Health
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.