Potassium disulphite
Catalog # KRAE109962
Supplier: KRAEMER MARTIN
CAS Number:
Specifications
- Pk:1
- Seq:0001
Specifications
About this item
:
:222.33 g/mol
Merck Index:13,07729
Melting Pt:190 °C
Density:1.2 g/cm³ (20 °C)
EINECS:240-795-3
:GHS05
:GHS07
CAS Number:16731-55-8
Formula:K₂O₅S₂
MDL Number:MFCD00167605