Thiophene-2-carbaldehyde, yellow liquid
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:98-03-3
:
Formula:C₅H₄OS
MDL Number:MFCD00005429
:112.15 g/mol
Boiling Pt:198 °C (1013 hPa)
Melting Pt:? 10 °C
Density:1.2 g/cm³ (25 °C)
Flash Pt:77 °C (lit.)
EINECS:202-629-8
:GHS07
Storage Temperature:Refrigerator