

Nitric acid 65%
Catalog # HONE17818.2500
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pk:2,5 L
- Seq:5096
Specifications
About this item
:
:63.01 g/mol
Merck Index:13,06608
Boiling Pt:120.5 °C (1013 hPa)
Melting Pt:–42 °C
Density:1.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
:GHS03
:GHS05
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349