2-Bromothiazole
Glass bottle for solids, 25 g
Catalog # FLUO021802-25G
Marketsource
Supplier: FLUOROCHEM
Jump to:
Product Details & Documents
- CAS Number:3034-53-5
- Formula:C₃H₂BrNS
- MDL Number:MFCD00005316
- Molecular Weight:164.03 g/mol
- Boiling Pt:171 °C (1013 hPa)
- Density:1,82 g/cm³ (20 °C)
- EINECS:221-229-4
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



