Order Entry
Germany
Orders LinkContactUsLinkComponent
3-(2-Chlorophenyl)propionic acid

3-(2-Chlorophenyl)propionic acid

Glass bottle for solids, 5 g
Artikelnummer FLUO018433-5G
Marketsource
Hersteller:   FLUOROCHEM
3-(2-Chlorophenyl)propionic acid
Artikelnummer FLUO018433-5G
Hersteller:  FLUOROCHEM
Marketsource
CAS-Nummer:  1643-28-3
3-(2-Chlorophenyl)propionic acid

Glass bottle for solids, 5 g

CAS-Nummer:  1643-28-3
Preis
52,50 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  3-(2-Chlorophenyl)propanoic acid
  • CAS Number:1643-28-3
  • Formula:C₉H₉ClO₂
  • MDL Number:MFCD00016547
  • Molecular Weight:184.62 g/mol
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.