Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(4-Methoxyphenyl)-2-thiourea
1-(4-Methoxyphenyl)-2-thiourea
1-(4-Methoxyphenyl)-2-thiourea
Toxic
1-(4-Methoxyphenyl)-2-thiourea
Supplier:
FLUOROCHEM
Certificates
Expand image
Toxic
1-(4-Methoxyphenyl)-2-thiourea
Supplier:
FLUOROCHEM
About this item
CAS Number:
2293-07-4
:
Formula:
(C₆H₄(OCH₃))NC(S)NH₂
MDL Number:
MFCD00004936
:
182.25 g/mol
EINECS:
218-931-8
UN:
2811
ADR:
6.1,III
:
GHS06