Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,3,3-Trifluoropropionaldehyde
3,3,3-Trifluoropropionaldehyde
3,3,3-Trifluoropropionaldehyde
3,3,3-Trifluoropropionaldehyde
Supplier:
FLUOROCHEM
Certificates
Expand image
3,3,3-Trifluoropropionaldehyde
Supplier:
FLUOROCHEM
About this item
CAS Number:
460-40-2
:
Formula:
C₃H₃F₃O
MDL Number:
MFCD00156074
:
112.05 g/mol
UN:
1988
ADR:
3,III