Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-(4-Fluorophenyl)propionic acid
3-(4-Fluorophenyl)propionic acid
3-(4-Fluorophenyl)propionic acid
3-(4-Fluorophenyl)propionic acid
FLUO006340-5G
:
FLUOROCHEM
:
3-(4-Fluorophenyl)propionic acid
FLUO006340-5G
:
FLUOROCHEM
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
168.17 g/mol
:
459-31-4
:
C₉H₉FO₂
:
MFCD00060327