3-Iodo-1-propanol
Catalog # FLUO002583-10G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:185.99 g/mol
Boiling Pt:201 °C (1013 hPa)
Melting Pt:–31 °C
Density:1,942 g/cm³ (25 °C)
EINECS:210-995-5
CAS Number:627-32-7
Formula:C₃H₇IO
MDL Number:MFCD00060329