Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2',4'-Difluoropropiophenone
2',4'-Difluoropropiophenone
2',4'-Difluoropropiophenone
2',4'-Difluoropropiophenone
Supplier:
FLUOROCHEM
Certificates
Expand image
2',4'-Difluoropropiophenone
Supplier:
FLUOROCHEM
About this item
CAS Number:
85068-30-0
:
Formula:
C₉H₈F₂O
MDL Number:
MFCD00015507
:
170.16 g/mol
Boiling Pt:
52 °C (1013 hPa)
Density:
1,193 g/cm³ (20 °C)
EINECS:
285-291-4