Order Entry
Germany
ContactUsLinkComponent
Dibenzothiophene
Dibenzothiophene
Catalog # EHERC20727000
Supplier:  EHRENSTORFER
CAS Number:  
Dibenzothiophene
Catalog # EHERC20727000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :184.26 g/mol
Boiling Pt:332 °C (1013 hPa)
Melting Pt:95…98 °C
Density:1,1295 g/cm³ (125 °C)
Flash Pt:170 °C
EINECS:205-072-9
CAS Number:132-65-0
Formula:C₁₂H₈S
MDL Number:MFCD00004969